Learn More
Apexbio Technology LLC Harmine, 500mg CAS# 442-51-3
SDP
Supplier:  Apexbio Technology LLC B8316500
Encompass_Preferred
Harmine, also known as telepathine, is an alkaloid of the -carboline family that regulates PPAR expression through inhibition of the Wnt signaling pathway. It also selectively binds to MAO-A and reversibly inhibits monoamine oxidase A (MAO-A) but not the variant MAO-B[3].Harmine has a severe cytotoxicity in hepatoma cells and holds great promise as an oncologic drug for anticancer treatment. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8253