Learn More
Selleck Chemical LLC Harmine hydrochloride S3817-1g
SDP
Catalog No. 503109047
Encompass_Preferred
Harmine (Telepathine) a fluorescent harmala alkaloid belonging to the beta-carboline family of compounds is a highly cell-permeant and competitive inhibitor of ATP binding to the kinase pocket of DYRK1A with about 60-fold higher IC50 value for DYRK2 Harmine also inhibits monoamine oxidases (MAOs) PPAR and cdc-like kinases (CLKs) Harmine inhibits 5-HT2A serotonin receptor with Ki of 397 nM
Catalog No. 50-310-9047 Supplier Selleck Chemical LLC Supplier No. S38171G
Harmine (Telepathine) a fluorescent harmala alkaloid belonging to the beta-carboline family of compounds is a highly cell-permeant and competitive inhibitor of ATP binding to the kinase pocket of DYRK1A with about 60-fold higher IC50 value for DYRK2 Harmine also inhibits monoamine oxidases (MAOs) PPAR and cdc-like kinases (CLKs) Harmine inhibits 5-HT2A serotonin receptor with Ki of 397 nM