Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmol Hydrochloride Dihydrate 98%, Thermo Scientific™

Catalog No. AC152270010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC152270010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152270010 Supplier Thermo Scientific Chemicals Supplier No. 152270010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 149022-16-2
Infrared Spectrum Authentic
Molecular Formula C12H15ClN2O3
Quantity 1g
MDL Number MFCD00150058
Synonym harmol hydrochloride dihydrate, 1-methyl-9h-pyrido 3,4-b indol-7-ol hydrochloride dihydrate, harmol hydrochloride hydrate, harmol dihydrate hydrochloride, c12h10n2o.hcl.2h2o, 1-methyl-2,9-dihydropyrido 3,4-b indol-7-one dihydrate hydrochloride
InChI Key OITPZRQHDRVLMO-UHFFFAOYSA-N
SMILES CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1.O.O.Cl
IUPAC Name 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one;dihydrate;hydrochloride
Molecular Weight (g/mol) 270.713
PubChem CID 2723830
Formula Weight 270.72
Percent Purity ≥97.5%
Chemical Name or Material Harmol hydrochloride dihydrate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.