Description
- Ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color
- Emulsified pigments are blended with an aqueous base to provide a consistent and high-quality product every time
- Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
- Dries in 3 minutes, improving laboratory workflow at the grossing station
- Non-flammable
- Available in round neck, induction sealed bottles with a standard flip top lid
- Includes one bottle of each color: Red, Yellow, Green, Blue, and Black
Specifications
Specifications
| Capacity | 2 oz. |
| Product Type | Tissue Marking Dye |
| Quantity | 5 Bottles |
| Form | Liquid |