Learn More
Mopec™ Harmony Tissue Marking Dye Kit with Stand
SDP
Catalog No. 22445842 Shop All Mopec Products
Change view
Click to view available options
Quantity:
7 Bottles
Catalog No. Quantity
22-445-842 7 Bottles
1 options
Catalog No. 22-445-842 Supplier Mopec™ Supplier No. BG40360S
Add to Cart
Add to Cart
Mopec™’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in 9 colors, with a standardized viscosity over the range.
  • Ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color
  • Emulsified pigments are blended with an aqueous base to provide a consistent and high-quality product every time
  • Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
  • Dries in 3 minutes, improving laboratory workflow at the grossing station
  • Non-flammable
  • Available in round neck, induction sealed bottles with a standard flip top lid
  • Includes one bottle of each color: Red, Yellow, Green, Blue, and Black
Specifications
Capacity 2 oz.
Product Type Tissue Marking Dye
Quantity 7 Bottles
Solution Type Liquid Ink
Form Liquid