Description
- Available in 9 colors, with a standardized viscosity over the range
- Ensures there is less variation in viscosity from one color to another as compared to competitor products
- Allows easy marking of both small and large specimens with any chosen color
- Emulsified pigments are blended with an aqueous base to provide a consistent and high-quality product every time
- Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
- Dries in 3 minutes, improving laboratory workflow at the grossing station
- Non-flammable
- Available in round neck, induction sealed bottles with a standard flip top lid
- Includes one 15 mL bottle of each color: Magenta, Red, Orange, Yellow, Green, Teal, Blue, Violet, and Black, and a small sample of Mopec™ Melody Shielding Spray and Mopec™ Amplify Setting Solution
Specifications
Specifications
| Product Type | Tissue Marking Dye |
| Quantity | 1 Bottle |
| Form | Liquid |