Learn More
Electron Microscopy Sciences Harms Micro Suturing Forceps, Curved
SDP
Catalog No. 5030216
Encompass_Preferred
These forceps are delicate with a 6 mm parallel platform. They are 4" long.
Catalog No. 50-302-16 Supplier Electron Microscopy Sciences Supplier No. 786144C
These forceps are delicate with a 6 mm parallel platform. They are 4" long.