Learn More
Instech Covance Infusion Harness™ for mice with .062in ID 12in (30cm) spring tether, non-sterile. For tethered infusion or sampling with mice.
SDP
Catalog No. 5081015
Encompass
The Instech CIH62 is a Covance Infusion Harness for mice designed to protect externalized catheters and provide stable tethering during infusion or sampling. It includes a molded elastomer saddle with a vented dome, a .062in ID 12in stainless‑steel spring tether for torque transmission, and adjustable belly bands to ensure a secure, comfortable fit throughout the study.
Features
  • .062in ID 12in stainless‑steel spring tether
  • Molded elastomer saddle with protective vented dome
  • Adjustable belly bands for proper fit and reduced stress
  • Supports infusion or blood sampling in mice
  • Designed to connect directly to standard Instech swivels
Catalog No. 50-810-15 Supplier Instech Supplier No. CIH62
The Instech CIH62 is a Covance Infusion Harness for mice designed to protect externalized catheters and provide stable tethering during infusion or sampling. It includes a molded elastomer saddle with a vented dome, a .062in ID 12in stainless‑steel spring tether for torque transmission, and adjustable belly bands to ensure a secure, comfortable fit throughout the study.
Features
  • .062in ID 12in stainless‑steel spring tether
  • Molded elastomer saddle with protective vented dome
  • Adjustable belly bands for proper fit and reduced stress
  • Supports infusion or blood sampling in mice
  • Designed to connect directly to standard Instech swivels