Learn More
Instech Covance Infusion Harness™ for mice, harness only, non-sterile. For tethered infusion or sampling with mice.
SDP
Catalog No. 5081017
Encompass
The Instech CIH62AB is a Covance Infusion Harness for mice designed to protect externalized catheters and provide stable tethering during infusion or sampling. It features a molded elastomer saddle with a vented dome, a .062in ID 12in stainless‑steel spring tether to transmit torque to the swivel, and adjustable belly bands that ensure a secure yet comfortable fit as the animal grows.
Features
  • .062in ID 12in stainless‑steel spring tether
  • Molded elastomer saddle with vented protective dome
  • Adjustable belly bands for proper fit and reduced stress
  • Supports infusion or blood sampling in tethered mice
  • Designed to connect directly to standard Instech swivels
Catalog No. 50-810-17 Supplier Instech Supplier No. CIH62AB
The Instech CIH62AB is a Covance Infusion Harness for mice designed to protect externalized catheters and provide stable tethering during infusion or sampling. It features a molded elastomer saddle with a vented dome, a .062in ID 12in stainless‑steel spring tether to transmit torque to the swivel, and adjustable belly bands that ensure a secure yet comfortable fit as the animal grows.
Features
  • .062in ID 12in stainless‑steel spring tether
  • Molded elastomer saddle with vented protective dome
  • Adjustable belly bands for proper fit and reduced stress
  • Supports infusion or blood sampling in tethered mice
  • Designed to connect directly to standard Instech swivels