Learn More
Instech Covance Infusion Harness™ for rats with .090in ID 12in (30cm) spring tether, non-sterile. For infusion or sampling with rats.
SDP
Catalog No. 5081018
Encompass
The Instech CIH95 is a Covance Infusion Harness for rats designed to protect an externalized catheter during single‑channel infusion or sampling. It features a molded elastomer saddle with a vented dome, a 12in stainless‑steel spring tether to transmit torque to the swivel, and adjustable belly bands that secure the harness while allowing for animal growth.
Features
  • 12in spring tether (.090in ID) for catheter protection
  • Molded elastomer saddle with vented dome
  • Adjustable belly bands for proper fit and comfort
  • Supports single‑channel infusion or sampling
  • Designed to connect directly to a swivel via the spring tether
Catalog No. 50-810-18 Supplier Instech Supplier No. CIH95
 Restricted Item
Add to Cart
Add to Cart
The Instech CIH95 is a Covance Infusion Harness for rats designed to protect an externalized catheter during single‑channel infusion or sampling. It features a molded elastomer saddle with a vented dome, a 12in stainless‑steel spring tether to transmit torque to the swivel, and adjustable belly bands that secure the harness while allowing for animal growth.
Features
  • 12in spring tether (.090in ID) for catheter protection
  • Molded elastomer saddle with vented dome
  • Adjustable belly bands for proper fit and comfort
  • Supports single‑channel infusion or sampling
  • Designed to connect directly to a swivel via the spring tether