Learn More
Instech Covance Infusion Harness™ and Belly Band; Dual channel infusion/sampling for rats, harness only, no spring, .105in ID, non sterile.
SDP
Catalog No. 5081014
Encompass
The Instech CIH105AB is a Covance Infusion Harness for rats designed with a larger .105in ID to support dual‑catheter configurations during infusion or sampling. This non‑sterile harness‑only model mates with the PS105 spring tether and provides stable exteriorization of tubing for reliable long‑term studies.
Features
  • .105in ID sized to accommodate two catheters
  • Harness‑only design for flexible system setup
  • Compatible with PS105 spring tethers
  • Supports dual‑channel infusion or blood sampling
  • Adjustable fit for secure long‑term use
Catalog No. 50-810-14 Supplier Instech Supplier No. CIH105AB
The Instech CIH105AB is a Covance Infusion Harness for rats designed with a larger .105in ID to support dual‑catheter configurations during infusion or sampling. This non‑sterile harness‑only model mates with the PS105 spring tether and provides stable exteriorization of tubing for reliable long‑term studies.
Features
  • .105in ID sized to accommodate two catheters
  • Harness‑only design for flexible system setup
  • Compatible with PS105 spring tethers
  • Supports dual‑channel infusion or blood sampling
  • Adjustable fit for secure long‑term use