Learn More
Instech Covance Infusion Harness™ Dual channel infusion/sampling of rats, .105in ID, 12in (30cm) spring tether, non-sterile.
SDP
Catalog No. 5081013
Encompass
The Instech CIH105 is a Covance Infusion Harness for rats designed for dual‑channel infusion or sampling. Its larger .105in ID allows routing of two catheters, while the 12in stainless‑steel spring tether protects externalized tubing and transmits torque to the swivel. This non‑sterile harness provides a secure, adjustable fit for long‑term studies.
Features
  • .105in ID to accommodate two catheters
  • Includes 12in stainless‑steel spring tether
  • Supports dual‑channel infusion or blood sampling
  • Protective spring design prevents catheter damage
  • Adjustable belly bands for stable, comfortable fit
Catalog No. 50-810-13 Supplier Instech Supplier No. CIH105
 Restricted Item
Add to Cart
Add to Cart
The Instech CIH105 is a Covance Infusion Harness for rats designed for dual‑channel infusion or sampling. Its larger .105in ID allows routing of two catheters, while the 12in stainless‑steel spring tether protects externalized tubing and transmits torque to the swivel. This non‑sterile harness provides a secure, adjustable fit for long‑term studies.
Features
  • .105in ID to accommodate two catheters
  • Includes 12in stainless‑steel spring tether
  • Supports dual‑channel infusion or blood sampling
  • Protective spring design prevents catheter damage
  • Adjustable belly bands for stable, comfortable fit