Learn More
Instech Covance Infusion Harness™ for rats, harness only, non-sterile. Mates with PS95 spring tether. For infusion or sampling with rats.
SDP
Catalog No. 5081019
Encompass
The Instech CIH95AB is a Covance Infusion Harness for rats designed to securely hold an implanted catheter during infusion or sampling studies. It provides stable exteriorization without a spring and is built to mate with PS95 spring tethers, offering a reliable connection point for swivels and tubing in long‑term experimental setups.
Features
  • Harness-only design for rat infusion and sampling
  • Compatible with PS95 spring tethers for secure attachment
  • Provides stable exteriorization of catheters
  • Non-sterile, durable construction for repeated use
  • Suitable for long-term infusion or IV self-administration studies
Catalog No. 50-810-19 Supplier Instech Supplier No. CIH95AB
The Instech CIH95AB is a Covance Infusion Harness for rats designed to securely hold an implanted catheter during infusion or sampling studies. It provides stable exteriorization without a spring and is built to mate with PS95 spring tethers, offering a reliable connection point for swivels and tubing in long‑term experimental setups.
Features
  • Harness-only design for rat infusion and sampling
  • Compatible with PS95 spring tethers for secure attachment
  • Provides stable exteriorization of catheters
  • Non-sterile, durable construction for repeated use
  • Suitable for long-term infusion or IV self-administration studies