Learn More
Med Vet International Jorvet Walkabout Harness, Front, X-Large, J0456FF
Catalog No. 5000495360
Encompass_Preferred
The Jorvet extra large Walkabout Harness is one of the only options for paralyzed dogs or cats. With their quality of life in mind today's pet owners use the Walkabout Hanress when they want choices to allow their pets to walk again. The design and construction fo the Walkabout harness provides the balance and support necessary for your pet to lead a ahppier, healthier, and cleaner life. The front harness has a smooth semicircular neckline that allows your pet to breathe comfortably while simultaneously supporting his/her front end. This walkabout harness has a chest circumference 31in.-35in. Upper foreleg circumference 16in.-19in. Front Harness
Catalog No. 50-004-95360 Supplier Med Vet International Supplier No. JOR456FF
The Jorvet extra large Walkabout Harness is one of the only options for paralyzed dogs or cats. With their quality of life in mind today's pet owners use the Walkabout Hanress when they want choices to allow their pets to walk again. The design and construction fo the Walkabout harness provides the balance and support necessary for your pet to lead a ahppier, healthier, and cleaner life. The front harness has a smooth semicircular neckline that allows your pet to breathe comfortably while simultaneously supporting his/her front end. This walkabout harness has a chest circumference 31in.-35in. Upper foreleg circumference 16in.-19in. Front Harness