Learn More
MSC Miller Full Body Harness, H700, 420 lb, Universal, 1 ea
Catalog No. 502834656
Encompass
The H700 Full Body Harness provides personal fall protection with a lightweight, universal design. Made from durable polyester, it supports a 420 lb load capacity and features multiple D-rings for versatile attachment.
  • One-pull trauma relief step for suspension trauma relief
  • Configurable leg strap design for customizable comfort
  • Modular lightweight accessory straps for pouch attachment
  • Lighter design with multi-layered padding for breathability and venting
  • Removable shoulder and leg padding for easy cleaning
  • Water repellent webbing for durability and easy maintenance
  • Integrated SRL connector for easy attachment of personal SRLs
  • Meets OSHA and ANSI Z359.11-2021 standards
Catalog No. 50-283-4656 Supplier MSC Supplier No. 96780945
The H700 Full Body Harness provides personal fall protection with a lightweight, universal design. Made from durable polyester, it supports a 420 lb load capacity and features multiple D-rings for versatile attachment.
  • One-pull trauma relief step for suspension trauma relief
  • Configurable leg strap design for customizable comfort
  • Modular lightweight accessory straps for pouch attachment
  • Lighter design with multi-layered padding for breathability and venting
  • Removable shoulder and leg padding for easy cleaning
  • Water repellent webbing for durability and easy maintenance
  • Integrated SRL connector for easy attachment of personal SRLs
  • Meets OSHA and ANSI Z359.11-2021 standards