Learn More
MSC DBI-SALA Fall Protection Harness, 420 Lb, Vest Style, Large, Back D-Ring
Catalog No. 502834938
Encompass
This DBI-SALA Fall Protection Harness is designed for general purpose fall protection applications. It features a vest style with a 420 Lb load capacity and quick-connect buckles on both leg and chest straps for easy donning.
  • Personal fall harness protection type.
  • General purpose application.
  • Large size.
  • Back D-ring location.
  • Quick-connect leg strap buckle type.
  • Quick-connect chest strap buckle type.
Catalog No. 50-283-4938 Supplier MSC Supplier No. 13110317
This DBI-SALA Fall Protection Harness is designed for general purpose fall protection applications. It features a vest style with a 420 Lb load capacity and quick-connect buckles on both leg and chest straps for easy donning.
  • Personal fall harness protection type.
  • General purpose application.
  • Large size.
  • Back D-ring location.
  • Quick-connect leg strap buckle type.
  • Quick-connect chest strap buckle type.