Learn More
MSC 3M DBI-SALA Harness, Vest Style, Universal, Polyester, Back D-Ring
Catalog No. 502835456
Encompass
The Delta Vest Safety Harness combines over two decades of design and manufacturing expertise. It features a patented triangular no-tangle back pad design and is made with tough, water-repellent, and abrasion-resistant webbing. This universal vest-style harness is designed for quick and easy donning, ensuring worker safety and comfort.
  • No-tangle design for easy donning
  • Stand-up back D-ring for efficient dorsal connections
  • Lightweight quick-connect buckle leg straps
  • Repel polyester webbing for durability
  • Built-in lanyard keepers
  • Revolving vertical torso buckle for easy adjustment
  • Impact indicator for safety checks
  • Protected labels equipped with integrated RFID tag
  • Integrated suspension trauma straps
Catalog No. 50-283-5456 Supplier MSC Supplier No. 31613763
The Delta Vest Safety Harness combines over two decades of design and manufacturing expertise. It features a patented triangular no-tangle back pad design and is made with tough, water-repellent, and abrasion-resistant webbing. This universal vest-style harness is designed for quick and easy donning, ensuring worker safety and comfort.
  • No-tangle design for easy donning
  • Stand-up back D-ring for efficient dorsal connections
  • Lightweight quick-connect buckle leg straps
  • Repel polyester webbing for durability
  • Built-in lanyard keepers
  • Revolving vertical torso buckle for easy adjustment
  • Impact indicator for safety checks
  • Protected labels equipped with integrated RFID tag
  • Integrated suspension trauma straps