Learn More
MSC DBI-SALA Fall Protection Harness, 420 Lb, Vest Style, Medium
Catalog No. 502835186
Encompass
This fall protection harness is designed for general purpose use. It features a vest style with a medium size and a 420 lb load capacity. The harness includes quick-connect buckles for both leg and chest straps, and a D-ring located on the back for personal fall protection.
  • Vest style
  • Medium size
  • General purpose application
  • Back D-ring location
  • Quick-connect leg strap buckle type
  • Quick-connect chest strap buckle type
  • 420 Lb load capacity
  • Personal fall protection type
Catalog No. 50-283-5186 Supplier MSC Supplier No. 20791653
This fall protection harness is designed for general purpose use. It features a vest style with a medium size and a 420 lb load capacity. The harness includes quick-connect buckles for both leg and chest straps, and a D-ring located on the back for personal fall protection.
  • Vest style
  • Medium size
  • General purpose application
  • Back D-ring location
  • Quick-connect leg strap buckle type
  • Quick-connect chest strap buckle type
  • 420 Lb load capacity
  • Personal fall protection type