Learn More
MSC 3M DBI-SALA Harness, Vest-Style, Universal, Back D-ring, 420 Lb
Catalog No. 502835197
Encompass
This vest-style harness is designed for general purpose fall protection, featuring a universal size and an automatic stand-up back D-ring for efficient dorsal connections. It includes integrated suspension trauma straps and auto-resetting lanyard keepers for enhanced safety and convenience. Protected labels with an integrated RFID tag facilitate inventory and inspection management.
  • Automatic stand-up back D-ring for efficient dorsal connections
  • Integrated suspension trauma straps for user safety
  • Auto-resetting lanyard keepers for convenient snap hook and carabiner parking
  • Protected labels with an integrated RFID tag for inventory and inspection management
  • Bright yellow webbing and triangular no-tangle back pad design
  • Universal size
  • Vest style
  • 420 Lb load capacity
  • Tongue leg strap buckle type
Catalog No. 50-283-5197 Supplier MSC Supplier No. 82724352
This vest-style harness is designed for general purpose fall protection, featuring a universal size and an automatic stand-up back D-ring for efficient dorsal connections. It includes integrated suspension trauma straps and auto-resetting lanyard keepers for enhanced safety and convenience. Protected labels with an integrated RFID tag facilitate inventory and inspection management.
  • Automatic stand-up back D-ring for efficient dorsal connections
  • Integrated suspension trauma straps for user safety
  • Auto-resetting lanyard keepers for convenient snap hook and carabiner parking
  • Protected labels with an integrated RFID tag for inventory and inspection management
  • Bright yellow webbing and triangular no-tangle back pad design
  • Universal size
  • Vest style
  • 420 Lb load capacity
  • Tongue leg strap buckle type