Learn More
MSC DBI-SALA Fall Protection Harness, 420 Lb, Vest, X-Large, 1 ea
Catalog No. 502834574
Encompass
Fall protection harnesses are designed for general purpose use, featuring a vest style and quick-connect straps for both legs and chest.
  • Load capacity of 420 lb
  • Vest style
  • X-large size
  • Polyester material
  • Back D-ring location
  • Quick-connect leg strap buckle type
  • Quick-connect chest strap buckle type
  • Personal fall protection type
  • General purpose application
Catalog No. 50-283-4574 Supplier MSC Supplier No. 31613847
Fall protection harnesses are designed for general purpose use, featuring a vest style and quick-connect straps for both legs and chest.
  • Load capacity of 420 lb
  • Vest style
  • X-large size
  • Polyester material
  • Back D-ring location
  • Quick-connect leg strap buckle type
  • Quick-connect chest strap buckle type
  • Personal fall protection type
  • General purpose application