Learn More
Enzo Life Sciences Harpagoside (25mg). CAS: 19210-12-9
SDP
Supplier:  Enzo Life Sciences ALX350333M025
Encompass_Preferred
Anti-inflammatory and anti-diabetic compound. Suppresses LPS-induced inducible nitric oxide synthase (iNOS; NOS II) and cyclooxygenase-2 (COX-2) expression through inhibition of NF-κB activation. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8848