Learn More
MSC Harrington Hoist Manual Lever Hoist, 1500 lb, 15 ft, ASME B30.21
Catalog No. 5000514736
Encompass
This manual lever hoist is engineered for demanding lifting applications. It offers a 1500 lb capacity and a 15 ft maximum lift, with durable construction and precision engineering for reliable performance in various industrial settings.
  • Steel construction for enhanced durability and longevity.
  • Ergonomic rubber grip provides comfort and secure handling.
  • Nickel-plated chain offers improved resistance to corrosion and wear.
  • Low headroom design allows for versatile use in confined spaces.
  • Double reduction gears ensure smooth and efficient operation.
  • Complies with ASME B30.21 standards for safety and performance.
  • 1500 lb pull capacity for heavy-duty lifting tasks.
  • Minimum headroom of 11 inches.
  • Equipped with a hook mount type for easy installation.
Catalog No. 50-005-14736 Supplier MSC Supplier No. 93699122
This manual lever hoist is engineered for demanding lifting applications. It offers a 1500 lb capacity and a 15 ft maximum lift, with durable construction and precision engineering for reliable performance in various industrial settings.
  • Steel construction for enhanced durability and longevity.
  • Ergonomic rubber grip provides comfort and secure handling.
  • Nickel-plated chain offers improved resistance to corrosion and wear.
  • Low headroom design allows for versatile use in confined spaces.
  • Double reduction gears ensure smooth and efficient operation.
  • Complies with ASME B30.21 standards for safety and performance.
  • 1500 lb pull capacity for heavy-duty lifting tasks.
  • Minimum headroom of 11 inches.
  • Equipped with a hook mount type for easy installation.