Learn More
MSC Harrington Hoist Mini Lever Hoist, 500 lb, 15 ft Lift, ASME B30.21
Catalog No. 5000514112
Encompass
The LX Mini Lever Hoist is a compact, award-winning tool designed for precise load positioning. It features a robust aluminum housing, heat-treated gears, and a nickel-plated load chain, ensuring durability and performance in challenging environments.
  • Compact, toolbox-sized design.
  • Durable aluminum housing and heat-treated gears.
  • Abrasion and corrosion-resistant nickel-plated load chain.
  • Precise load positioning in tight or elevated spaces.
  • Lightweight body and low operating force.
  • Quick two-step free-chain and hook positioning.
  • Totally enclosed Weston-style load brake.
  • Complies with ANSI/ASME B30.21 and HST-3M standards.
Catalog No. 50-005-14112 Supplier MSC Supplier No. 39163845
The LX Mini Lever Hoist is a compact, award-winning tool designed for precise load positioning. It features a robust aluminum housing, heat-treated gears, and a nickel-plated load chain, ensuring durability and performance in challenging environments.
  • Compact, toolbox-sized design.
  • Durable aluminum housing and heat-treated gears.
  • Abrasion and corrosion-resistant nickel-plated load chain.
  • Precise load positioning in tight or elevated spaces.
  • Lightweight body and low operating force.
  • Quick two-step free-chain and hook positioning.
  • Totally enclosed Weston-style load brake.
  • Complies with ANSI/ASME B30.21 and HST-3M standards.