Learn More
Medchemexpress LLC HY-N0862 5mg , Harringtonine CAS:26833-85-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08625MG
Encompass_Preferred
HY-N0862 5mg Harringtonine CAS: 26833-85-2 Harringtonine is a natural Cephalotaxus alkaloid that inhibits protein synthesis. Harringtonine has anti-chikungunya virus (CHIKV) activities with an EC50 of 0.24 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9829