Learn More
Lkt Laboratories Harringtonine, ≥97%, 5MG
SDP
Supplier:  Lkt Laboratories H01695MG
Encompass_Preferred
Cephalotaxine alkaloid originally found in Cephalotaxus; ribosomal protein synthesis inhibitor. CAS: 26833-85-2. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4855