Learn More
Grainger HARRIS Specialty Gas Regulator, Single Stage, CGA 580
Catalog No. NC2747886
Encompass_Preferred
The Harris Specialty Gas Regulator is a single stage regulator designed for specialty gases. It has a CGA 580 inlet and outlet connection and is made of brass construction. The regulator has a pressure range of 0-100 PSI and features a pressure relief valve, outlet pressure gauge, and flow capacity of 60 SCFH air. The regulator body measures 3-1/2" diameter x 5-3/4" high. This regulator is suitable for use with gases like hydrogen, helium, argon, and nitrogen. It provides accurate pressure regulation for specialty gas cylinder applications.
Catalog No. NC2747886 Supplier Grainger Supplier No. 30AA57
The Harris Specialty Gas Regulator is a single stage regulator designed for specialty gases. It has a CGA 580 inlet and outlet connection and is made of brass construction. The regulator has a pressure range of 0-100 PSI and features a pressure relief valve, outlet pressure gauge, and flow capacity of 60 SCFH air. The regulator body measures 3-1/2" diameter x 5-3/4" high. This regulator is suitable for use with gases like hydrogen, helium, argon, and nitrogen. It provides accurate pressure regulation for specialty gas cylinder applications.