Learn More
Anatech Ltd HARRIS HEMATOXYLIN 1GAL
Catalog No. NC9520196
Encompass_Preferred
HARRIS HEMATOXYLIN, 1 Gallon, General use:A blue nuclear stain in histopathology, Product description:Aqueous solution of hematoxylin, Component:Acetic acid; Aluminum ammonium sulfate; Harris (C.I. 75290)
Catalog No. NC9520196 Supplier Anatech Ltd Supplier No. 842
HARRIS HEMATOXYLIN, 1 Gallon, General use:A blue nuclear stain in histopathology, Product description:Aqueous solution of hematoxylin, Component:Acetic acid; Aluminum ammonium sulfate; Harris (C.I. 75290)