Learn More
Electron Microscopy Sciences Harris Hematoxylin 500 ML
SDP
Catalog No. 5031837
Encompass_Preferred
Used in Herxheimer's Technique for staining fat and cholesterol. References: Clark, G., Ed.: Staining Procedures; 3rd Ed.; Baltimore: Williams and Wilkins Co., c. 1973, p. 143 Conn's, H.J.: Biological Stains; 8th Ed., Baltimore: Williams and Wilkins Co., c. 1969, p.100
Catalog No. 50-318-37 Supplier Electron Microscopy Sciences Supplier No. 2650002
Used in Herxheimer's Technique for staining fat and cholesterol. References: Clark, G., Ed.: Staining Procedures; 3rd Ed.; Baltimore: Williams and Wilkins Co., c. 1973, p. 143 Conn's, H.J.: Biological Stains; 8th Ed., Baltimore: Williams and Wilkins Co., c. 1969, p.100