Learn More
StatLab™ Harris Hematoxylin Stain
GSA_VASDP
Catalog No. STLSL9016
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL9016 16 oz.
STLSL901 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL9016 Supplier StatLab™ Supplier No. SL9016
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Modified traditional formulation of Harris Hematoxylin.
  • Contains no mercury
  • Generally used as a regressive stain, but may also be used in progressive staining protocols
  • Produces rapid and distinct nuclear detail
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 16 oz.
Product Type Hematoxylin