Learn More
Electron Microscopy Sciences Harris Hematoxylin w/o Mercury 500 ML
SDP
Catalog No. 5026448
Encompass_Preferred
Harris' Hematoxylin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used to selectivity stain for nuclei, fat, and lipids.
Catalog No. 50-264-48 Supplier Electron Microscopy Sciences Supplier No. 2604120
Harris' Hematoxylin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used to selectivity stain for nuclei, fat, and lipids.