Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Flagging Tape

Catalog No. 19040023
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
White
Length (English):
150 ft.
300 ft.
Thickness:
2 mil
4 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Thickness
19-040-023 Blue 150 ft. 4 mil
19-811-853 White 300 ft. 2 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-040-023 Supplier Harris Industries Supplier No. VST21
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-adhesive tapes available in standard or fluorescent colors

  • 3cm (1.19 in.) width
  • 3.81cm (1.5 in.) core
  • Non-adhesive
  • Vinyl with monomeric flexible PVC film
  • Taffeta embossing pattern

Specifications

Length (English) 150 ft.
For Use With (Application) Marking hazards, trees, danger areas, flagging boundaries and survey markers
Type Flagging Tape
Color Blue
Material PVC
Length (Metric) 45.72 m
Tensile Strength 2620 to 2990 psi
Thickness 4 mil
Width (English) 1.19 in.
Width (Metric) 3 cm
Adhesive Type Non-adhesive
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.