Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Flagging Tape

Catalog No. 19811853
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
White
Length (English):
150 ft.
300 ft.
Thickness:
2 mil
4 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Thickness
19-811-853 White 300 ft. 2 mil
19-040-023 Blue 150 ft. 4 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-811-853 Supplier Harris Industries Supplier No. VST3
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-adhesive tapes available in standard or fluorescent colors

  • 3cm (1.19 in.) width
  • 3.81cm (1.5 in.) core
  • Non-adhesive
  • Vinyl with monomeric flexible PVC film
  • Taffeta embossing pattern

Specifications

For Use With (Application) Marking hazards, trees, danger areas, flagging boundaries and survey markers
Width (Metric) 3 cm
Thickness 2 mil
Width (English) 1.19 in.
Length (English) 300 ft.
Length (Metric) 91.4 m
Tensile Strength 2620 to 2990 psi
Adhesive Type Non-adhesive
Color White
Material PVC
Type Flagging Tape
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.