Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries General-Purpose Masking Tape

Catalog No. 19059429
Encompass
Encompass_Preferred
Change view
Click to view available options
Width (English):
2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
19-059429 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-059429 Supplier Harris Industries Supplier No. GP002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Adhesive Type Rubber
Format Tape
Product Type Masking Tape
For Use With (Application) Used for masking, bundling, sealing, splicing and surface protection
Quantity 1 Ea.
Length (English) 60 yd.
Length (Metric) 54.8 m
Width (English) 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.