Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Nonslip Tapes

Catalog No. 19811819
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-811-819 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-811-819 Supplier Harris Industries Supplier No. AS3
Add to Cart
Add to Cart

Specifications

Description Surface Mount
Adhesive Type Self-adhesive
Color Black
Format Tape
Product Type Nonslip Tape
For Use With (Application) Used in high traffic areas where slippery or dangerous surfaces may cause accidents
Quantity 1 Ea.
Length (English) 60 ft.
Length (Metric) 18.2 m
Width (English) 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.