Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Pennant Lines

Catalog No. 19811844
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-811-844 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-811-844 Supplier Harris Industries Supplier No. PPL49
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Color Black, Yellow
Product Type Printed Pennant Line
Display Text Danger
For Use With (Application) Traffic control, marking of hazardous areas or for open-houses, sales events or any place of high visibility
Quantity 1 Ea.
Size (English) 9 x 12 in.
Size (Metric) 22.8 x 30.4 cm
Length (English) 60 ft.
Length (Metric) 18.2 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.