Learn More
Sigma Aldrich Fine Chemicals Biosciences Harris Uni Core I.D. 1.2 m
Catalog No. 502904038
Encompass
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction collection and storage in one operation. Suitably samples thin and soft substrates including filter paper gels and dried blood on cards. Units can be autoclaved at 250C for limited re-use.
Catalog No. 50-290-4038 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WHAWB100074
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction collection and storage in one operation. Suitably samples thin and soft substrates including filter paper gels and dried blood on cards. Units can be autoclaved at 250C for limited re-use.