Learn More
Ted Pella Inc HARRIS UNI-CORE, TIP ID 3.0MM
SDP
Supplier:  Ted Pella Inc 15078
Encompass
The Harris Uni-Core consists of a razor sharp stainless steel cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates.
Catalog No. NC0917983