Learn More
Aviva Systems Biology Corp HARS2 Antibody - C-terminal region (ARP62671_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HARS2. It was validated on Western Blot. . Alternate names: HARSL, HARSR, HO3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-733