Learn More
Hach Company Hart Network Comm. Mod. SC200
Supplier:  Hach Company 9328100
Encompass
Hart Network Communication Module for SC200 Universal ControllerThe HART communication module for the SC200 is a user installed module that allows for communication via HART from the controller See manual for additional connection information
Catalog No. 50-002-08102