Learn More
Med Vet International Miltex Vantage Hartman-Noyes Alligator Forceps, 3.25in., Serrated Jaws, Extra Delicate Pattern
Catalog No. 502830285
Encompass_Preferred
The Miltex Vantage V919-434A Hartman-Noyes alligator ear forceps are made from high quality Japanese stainless steel and measure 3.25" in length. They have a non-locking finger ring handle and serrated alligator jaws with an extra delicate pattern.
Catalog No. 50-283-0285 Supplier Med Vet International Supplier No. MILV919434A
The Miltex Vantage V919-434A Hartman-Noyes alligator ear forceps are made from high quality Japanese stainless steel and measure 3.25" in length. They have a non-locking finger ring handle and serrated alligator jaws with an extra delicate pattern.