Learn More
Roboz Surgical Hartman Mosquito Forceps; Curved; 4" Length, 0.8 mm Tip Diameter
SDP
Catalog No. NC9199225
Encompass
Hartman Mosquito Forceps; Curved; 4 in. Length, 0.8mm tip diameter, 1.9mm jaw width @ 5mm from the tip. The curved version of our shortest hemostats is 4 in. long. The tip diameter is 0.8mm, the jaw width is 1.9mm measured at 5mm in from the tip.
Catalog No. NC9199225 Supplier Roboz Surgical Supplier No. RS7101
Hartman Mosquito Forceps; Curved; 4 in. Length, 0.8mm tip diameter, 1.9mm jaw width @ 5mm from the tip. The curved version of our shortest hemostats is 4 in. long. The tip diameter is 0.8mm, the jaw width is 1.9mm measured at 5mm in from the tip.