Learn More
Med Vet International Miltex Vantage Hartman Mosquito Hemostatic Forceps, 3.5in, Curved
Catalog No. 502559426
Encompass_Preferred
The Miltex Vantage V97-26 Hartman mosquito forceps are made from high quality Japanese stainless steel and measure 3.5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.
Catalog No. 50-255-9426 Supplier Med Vet International Supplier No. MILV9726
The Miltex Vantage V97-26 Hartman mosquito forceps are made from high quality Japanese stainless steel and measure 3.5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.