Learn More
Roboz Surgical HARTMAN MOSQ STRAIGHT
SDP
Catalog No. NC9393758
Encompass
Hartman Mosquito Forceps; Straight; 4 in. Length, 0.8mm tip diameter, 1.9mm jaw width @ 5mm from the tip. The Hartman Mosquito Forceps are our shortest hemostats with only 4 in. length. The tip diameter is 0.8 mm. The tip jaw width is 1.9 mm at 5 mm fr
Catalog No. NC9393758 Supplier Roboz Surgical Supplier No. RS7100
Hartman Mosquito Forceps; Straight; 4 in. Length, 0.8mm tip diameter, 1.9mm jaw width @ 5mm from the tip. The Hartman Mosquito Forceps are our shortest hemostats with only 4 in. length. The tip diameter is 0.8 mm. The tip jaw width is 1.9 mm at 5 mm fr