Learn More
Stoelting™ Hartman Mosquito Forceps
SDP
Catalog No. 10000471 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
4 in.
Model:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-000-471 4 in. Curved
10-000-470 4 in. Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-471 Supplier Stoelting™ Supplier No. 5211001P
Add to Cart
Add to Cart
Stoelting™ Hartman Mosquito Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
  • Delicate hemostats have thin jaws with fine serrations
Specifications
For Use With (Application) Neuroscience
Length (English) 4 in.
Model Curved