Learn More
World Precision Instrument Hartman Mosquito Forceps, 9cm, Straight, 12-pack, Disposable
SDP
Catalog No. 502539442
Encompass
Hartman Mosquito Forceps, also called Hartmann mosquito hemostatic forceps, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Hartman Mosquito Forceps have a larger tip and deeper serrations than Halsted Mosquito Forceps. They are designed for clamping larger blood vessels. 
Catalog No. 50-253-9442 Supplier World Precision Instrument Supplier No. 50372812
Hartman Mosquito Forceps, also called Hartmann mosquito hemostatic forceps, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Hartman Mosquito Forceps have a larger tip and deeper serrations than Halsted Mosquito Forceps. They are designed for clamping larger blood vessels. 