Learn More
Sklar Instruments Forceps, Surgi-OR Hartmann Mosquito Forceps, Sklar 3-1/2 in., Curved, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126500 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Curved, Serrated, Stainless Steel, EA. Surgi-OR Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6500 Supplier Sklar Instruments Supplier No. 95421
Forceps, Surgi-OR Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Curved, Serrated, Stainless Steel, EA. Surgi-OR Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Lifetime warranty against all manufacturer defects.