Learn More
Stoelting™ Hartman Rongeur
SDP
Catalog No. 10001802 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Strong Curve
Catalog No. Model
10-001-802 Strong Curve
1 options
Catalog No. 10-001-802 Supplier Stoelting™ Supplier No. 5216302
Add to Cart
Add to Cart
Rongeurs feature a sharp-edged, scoop-shaped tip, used for gouging out bone or opening a window in bone, often in the skull. It is used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
Length (English) 5.75 in.
Model Strong Curve