Learn More
Hartman's Fixative, Sigma-Aldrich
Catalog No. 01259032 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
01-259-032 500 mL
1 options
Catalog No. 01-259-032 Supplier MilliporeSigma Supelco Supplier No. H0290500ML
Add to Cart
Add to Cart
Hartman's fixative, which is also known as Davidson's Fixative, contains acetic acid, alcohol, and formalin. It is widely used for the preservation of eye, bone marrow, breast, testis, and other tissues for histologic examination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixative
Quantity 500 mL
Solution Type Fixative
Form Liquid