Learn More
Sklar Instruments Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. EA/1.
SDP
Catalog No. 501437392 Shop All Sklar Instruments Products
Encompass_Preferred
Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. This product is straight with serrated tips and a length of 3-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7392 Supplier Sklar Instruments Supplier No. 171235
Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. This product is straight with serrated tips and a length of 3-1/2 inches.