Learn More
Sklar Instruments Sklarlite Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Curved, Serrated, Extra Light (SklarLite), Stainless Steel. EA/1.
SDP
Catalog No. 501437952 Shop All Sklar Instruments Products
Encompass_Preferred
Sklarlite Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Curved, Serrated, Extra Light (SklarLite), Stainless Steel. Sklarlite Hartmann Mosquito forceps are a lighter version of standard mosquito forceps. This lightweight modification provides the surgeon with greater control and less fatigue. Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. This product is curved with serrated tips and a length of 3-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7952 Supplier Sklar Instruments Supplier No. 232101
Sklarlite Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Curved, Serrated, Extra Light (SklarLite), Stainless Steel. Sklarlite Hartmann Mosquito forceps are a lighter version of standard mosquito forceps. This lightweight modification provides the surgeon with greater control and less fatigue. Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. This product is curved with serrated tips and a length of 3-1/2 inches.